4-Cyano-1-butyne 97%

Danger

AC45559-0050 AC45559-0010
103534-244EA 40.4 USD
103534-244 103534-246
4-Cyano-1-butyne 97%
4-Cyano-1-butyne
Formula: C₅H₅N
Boiling Pt: 79 °C (39 mmHg)
Density: 0.9063 g/cm³
Flash Pt: 65.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041563
CAS Number: 19596-07-7
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4380 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR