Lithium thiophenoxide 0.6M in THF, AcroSeal™

Danger

Synonyms: Lithium thiophenolate

AC43350-1000
103533-416EA 186.6 USD
103533-416
Lithium thiophenoxide 0.6M in THF, AcroSeal™
Lithium thiophenoxide
Formula: C₆H₅LiS
Boiling Pt: 67 °C (760 mmHg)
Density: 0.934 g/cm³
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00192253
CAS Number: 2973-86-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to light yellow Solution
Titration with HClO4 0.55 to 0.65 M
Melting point 45 to 50 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR