(±)-Boc-α-phosphonoglycine trimethyl ester 97%

Danger

Synonyms: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

AC43921-0050 AC43921-0010
103533-632EA 52.3 USD
103533-632 103533-634
(±)-Boc-α-phosphonoglycine trimethyl ester 97%
(±)-Boc-α-phosphonoglycine trimethyl ester
Formula: C₁₀H₂₀NO₇P
MW: 297.24 g/mol
Melting Pt: 69.7…71.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02178893
CAS Number: 89524-98-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR