Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥97%

Supplier: ACROS ORGANICS MS

Warning

AC10026-0050 AC10026-0010
103529-992EA 75 USD InStock
103529-992 103529-990
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone ≥97%
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone


Formula: C₁₂H₁₄ClN₃O
MW: 251,72 g/mol
Melting Pt: 228…231 °C
Storage Temperature: Ambient
MDL Number: MFCD02093793
CAS Number: 53786-28-0
EINECS: 258-771-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 224°C to 228°C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty