2-Chloroquinoxaline 97%

Warning

AC13619-0050 AC13619-0010
103530-958EA 24.6 USD
103530-958 103530-960
2-Chloroquinoxaline 97%
2-Chloroquinoxaline
Formula: C₈H₅ClN₂
MW: 164.5917 g/mol
Boiling Pt: 100 °C (1.4 mmHg)
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00043907
CAS Number: 1448-87-9
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to grey to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 47 to 51 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR