3-Amino-5-bromo-1H-indazole 95%

Warning

Synonyms: 5-Bromo-1H-indazol-3-amine

AC43576-0010 AC43576-0050
103533-486EA 116 USD
103533-486 103533-488
3-Amino-5-bromo-1H-indazole 95%
3-Amino-5-bromo-1H-indazole
Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Melting Pt: 237…238 °C
Storage Temperature: Ambient
MDL Number: MFCD03426696
CAS Number: 61272-71-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 178 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR