3-Ethynylthiophene 96%

Danger

AC43858-0050 AC43858-0010
103533-594EA 253 USD
103533-594 103533-592
3-Ethynylthiophene 96%
3-Ethynylthiophene
Formula: C₆H₄S
MW: 108.16 g/mol
Boiling Pt: 152…154 °C (760 mmHg)
Density: 1.098 g/cm³
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039973
CAS Number: 67237-53-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Liquid (May darken on storage)
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR