3-Amino-5-(trifluoromethyl)-1H-indazole 95%

Warning

AC43573-0010 AC43573-0050
103533-484EA 592.9 USD
103533-484 103533-482
3-Amino-5-(trifluoromethyl)-1H-indazole 95%
3-Amino-5-(trifluoromethyl)-1H-indazole
Formula: C₈H₆F₃N₃
Melting Pt: 134.5…135.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01719740
CAS Number: 2250-53-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 133 to 137 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR