1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Warning

Synonyms: 1-Isobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

AC43973-0010 AC43973-0050
103530-718EA 74 USD
103530-718 103530-720
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₃H₂₃BN₂O₂
MW: 250.1449 g/mol
Boiling Pt: 248…249 °C (760 mmHg)
Density: 1.003 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD05663857
CAS Number: 827614-66-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR