(3-Chlorophenyl)methanesulfonyl chloride 97%

Danger

Synonyms: 3-Chlorobenzylsulphonyl chloride3-Chlorobenzylsulfonyl chloride3-Chloro-α-toluenesulphonyl chloride

AC45448-0050 AC45448-0250 AC45448-0010
103534-210EA 63 USD
103534-210 103534-212 103534-208
(3-Chlorophenyl)methanesulfonyl chloride 97%
(3-Chlorophenyl)methanesulphonyl chloride
Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Melting Pt: 71…75 °C
Storage Temperature: Ambient
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 % As diethylamine derivative
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR