1H-Benzimidazole-4-carboxylic acid 97%

Warning

Synonyms: 1H-Benzimidazole-7-carboxylic acid

AC42951-0010 AC42951-0050
103533-242EA 1285 USD
103533-242 103533-240
1H-Benzimidazole-4-carboxylic acid 97%
1H-Benzimidazole-4-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01823426
CAS Number: 46006-36-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR