Benzyltriphenylphosphonium tetrahydroborate ≥95%
|
Danger
|
Synonyms:
Benzyltriphenylphosphonium borohydride
AC37637-0050
AC37637-0250
103532-704EA
135.4
USD
103532-704
103532-702
Benzyltriphenylphosphonium tetrahydroborate ≥95%
Benzyltriphenylphosphonium tetrahydroborate
Formula:
C₂₅H₂₆BP Melting Pt: 157 …163 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00800636 CAS Number: 18117-29-8 UN: 3131 ADR: 4.3,II |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 157°C to 163°C |
Titration with HClO4 | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...