1-(Trimethylsilyl)benzotriazole 97%

Danger

Synonyms: 1-(Trimethylsilyl)-1H-benzotriazole

AC42626-0050 AC42626-0010
103533-154EA 52.5 USD
103533-154 103533-152
1-(Trimethylsilyl)benzotriazole 97%
1-(Trimethylsilyl)benzotriazole
Formula: C₉H₁₃N₃Si
Boiling Pt: 125…126 °C (3 mmHg)
Density: 1.054 g/cm³
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00216577
CAS Number: 43183-36-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR