(S)-(-)-1-Boc-3-aminopyrrolidine 95%

Danger

Synonyms: (S)-1-Boc-3-Aminopyrrolidine(S)-(-)-1-BOC-3-aminopyrrolidinetert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

AC43545-0010 AC43545-0050
103533-470EA 344 USD
103533-470 103533-468
(S)-(-)-1-Boc-3-aminopyrrolidine 95%
(S)-(-)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Boiling Pt: 120…125 °C (5Torr)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD03419271
CAS Number: 147081-44-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Specific optical rotation -1.5° to -3.0° (20 °C, 589 nm) (C=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR