Benzamidoxime 97%

Danger

Synonyms: Benzamide oximeN'-Hydroxybenzimidamide

AC45328-0050 AC45328-0010
103534-134EA 97.4 USD
103534-134 103534-132
Benzamidoxime 97%
Benzamidoxime
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00031485
CAS Number: 613-92-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 73 to 79 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR