Phenethyltrichlorosilane 95%

Danger

AC43224-0010
103529-764EA 31.4 USD
103529-764
Phenethyltrichlorosilane 95%
Phenethyltrichlorosilane
Formula: C₈H₉Cl₃Si
Boiling Pt: 93…96 °C (3 mmHg)
Density: 1.24 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00039290
CAS Number: 940-41-0
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5175 to 1.5195 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR