2,3-Dibromo-3-phenylpropiophenone 98%

Danger

AC37508-0010 AC37508-0050
103529-610EA 14.5 USD
103529-610 103529-612
2,3-Dibromo-3-phenylpropiophenone 98%
2,3-Dibromo-3-phenylpropiophenone
Formula: C₁₅H₁₂Br₂O
Melting Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00017861
CAS Number: 611-91-6
EINECS: 210-282-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR