6-Chloronicotinoyl chloride 97%

Danger

AC44225-0250
103533-752EA 106.3 USD
103533-752
6-Chloronicotinoyl chloride 97%
6-Chloronicotinoyl chloride
Formula: C₆H₃Cl₂NO
Boiling Pt: 128…130 °C (23Torr)
Melting Pt: 48…50 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051775
CAS Number: 58757-38-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 47 to 51 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR