(S)-(+)-3-Amino-1-boc-piperidine 95%

Danger

Synonyms: (S)-(+)-3-Amino-1-BOC-piperidine(S)-tert-Butyl 3-aminopiperidine-1-carboxylate

AC43749-0010 AC43749-0050
103530-602EA 653 USD
103530-602 103530-600
(S)-(+)-3-Amino-1-boc-piperidine 95%
(S)-(+)-3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094718
CAS Number: 625471-18-3
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to yellow Gum or semi-solid
Infrared spectrum Conforms
GC ≥94.0 %
Specific optical rotation +26.5° to +30.5° (20 °C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR