Diclofenac sodium salt 98%

Danger

Synonyms: [o-(2,6-dichloroanilino) phenyl]acetic acid sodium saltSodium [o-[(2,6-dichlorophenyl) amino] phenyl] acetate2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium saltGP-458402-[(2,6-Dichlorophenyl)amino]-benzeneacetic acid monosodium salt

AC44525-0100
103529-858EA 59.9 USD
103529-858
Diclofenac sodium salt 98%
Diclofenac sodium salt
Formula: C₁₄H₁₀Cl₂NNaO₂
MW: 318.1 g/mol
Melting Pt: 283…285 °C
Storage Temperature: Ambient
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %
pH 6.5 to 7.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR