(Methylthio)acetaldehyde dimethyl acetal 98%

Danger

Synonyms: (Methylthio)acetaldehydedimethylacetal

AC43201-0100
103529-746EA 42 USD
103529-746
(Methylthio)acetaldehyde dimethyl acetal 98%
(Methylthio)acetaldehyde dimethyl acetal
Formula: C₅H₁₂O₂S
MW: 136.21 g/mol
Boiling Pt: 60 °C (13 mmHg)
Density: 1.021 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4550 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR