(S)-(-)-N,α-Dimethylbenzylamine 99+%

Danger

Synonyms: (S)-(-)-N-Methyl-1-phenylethylamine(S)-N,α-Dimethylbenzylamine(S)-(-)-N α-Dimethylbenzylamine(S)-(-)-alpha-Methyl-N-methylbenzylamine

AC39584-0050 AC39584-0010
103532-980EA 163 USD
103532-980 103532-978
(S)-(-)-N,α-Dimethylbenzylamine 99+%
(S)-(-)-N,α-Dimethylbenzylamine
Formula: C₉H₁₃N
MW: 135.2100 g/mol
Boiling Pt: 74…76 °C (11 mmHg)
Density: 0.924 g/cm³
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00067113
CAS Number: 19131-99-8
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5130 (20 °C, 589 nm)
GC ≥99.0 %
Specific optical rotation -63° to -67° (20 °C, 589 nm) (c=4, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR