Cyclopropanesulfonyl chloride 97%

Danger

Synonyms: CyclopropanesulphonylchlorideCyclopropanesulfonylchloride

AC43902-0010 AC43902-0050
103533-628EA 889 USD
103533-628 103533-626
Cyclopropanesulfonyl chloride 97%
Cyclopropanesulphonyl chloride
Formula: C₃H₅ClO₂S
MW: 140.59 g/mol
Boiling Pt: 60 °C (2Torr)
Density: 1.38 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01631933
CAS Number: 139631-62-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR