Thiophene-3-methylamine 97%

Danger

Synonyms: 3-Thienylmethylamine

AC45145-0100
103529-908EA 464.4 USD
103529-908
Thiophene-3-methylamine 97%
Thiophene-3-methylamine
Formula: C₅H₇NS
MW: 113.18
Boiling Pt: 102 °C (15 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD01529872
CAS Number: 27757-86-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR