Tris(trimethylsilyl)borate

Warning

Synonyms: Tris(trimethylsiloxy)borate

AC38124-1000 AC38124-0250
103532-784EA 345 USD
103532-784 103529-652
Tris(trimethylsilyl)borate
Tris(trimethylsilyl) borate
Formula: C₉H₂₇BO₃Si₃
MW: 278.376 g/mol
Boiling Pt: 183…185 °C (760 mmHg)
Density: 0.834 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00051588
CAS Number: 4325-85-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3870 (20 °C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR