Cyclopropylboronic acid pinacol ester 95%

Warning

Synonyms: 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

AC42859-0010 AC42859-0050
103533-184EA 78.9 USD
103533-184 103533-186
Cyclopropylboronic acid pinacol ester 95%
Cyclopropylboronic acid pinacol ester
Formula: C₉H₁₇BO₂
MW: 168.04 g/mol
Boiling Pt: 146 °C (760 mmHg)
Density: 0.922 g/cm³
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663847
CAS Number: 126689-01-8
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR