1-Pentylmagnesium chloride in THF, AcroSeal™

Danger

Synonyms: Pentylmagnesium chloride

AC43621-1000
103529-812EA 39.3 USD
103529-812
1-Pentylmagnesium chloride in THF, AcroSeal™
1-Pentylmagnesium chloride
Formula: C₅H₁₁ClMg
MW: 130.9
Boiling Pt: 65 °C (760 mmHg)
Density: 0.967 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00672089
CAS Number: 6393-56-2
UN: 3399
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Clear liquid
Titration Grignard 1.20 to 1.40 M
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR