4,5-Diaminopyrimidine 97%

Warning

Synonyms: Pyrimidine-4,5-diamine

AC11240-0010
103530-498EA 189 USD
103530-498
4,5-Diaminopyrimidine 97%
4,5-Diaminopyrimidine
Formula: C₄H₆N₄
MW: 110.1200 g/mol
Boiling Pt: 229 °C (32 mmHg)
Melting Pt: 200 °C
Storage Temperature: Ambient
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥97.5 %
Specific optical rotation +53° to +56° (22°C, 589 nm) (c=1, 1,2-dichlorethane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR