N-Methyl-1,3-propanediamine 99%

Danger

Synonyms: 3-Methylamino propylamineN-Methylpropane-1,3-diamine

AC15557-0025 AC15557-2500 AC15557-0010
103531-430EA 188 USD
103531-430 103531-432 103531-434
N-Methyl-1,3-propanediamine 99%
N-Methyl-1,3-propanediamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 139…141 °C (760 mmHg)
Melting Pt: –72 °C
Density: 0.84 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00008209
CAS Number: 6291-84-5
EINECS: 228-544-6
UN: 2922
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR