Cyclopropylacetic acid 97%

Danger

AC44378-0050 AC44378-0010
103533-830EA 65.2 USD
103533-830 103533-832
Cyclopropylacetic acid 97%
Cyclopropylacetic acid
Formula: C₅H₈O₂
Boiling Pt: 60…61 °C (29 mmHg)
Density: 1.14 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00041544
CAS Number: 5239-82-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR