2-Fluoropyrazine 98%

Danger

AC45696-0010
103534-318EA 57 USD
103534-318
2-Fluoropyrazine 98%
2-Fluoropyrazine
Formula: C₄H₃FN₂
MW: 98.0784 g/mol
Boiling Pt: 110 °C
Density: 1.213 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD03094084
CAS Number: 4949-13-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR