Tetrabutylammonium dihydrogen phosphate 0.4 M in acetonitrile, AcroSeal™

Danger

Synonyms: Tetra-n-butylammonium phosphate monobasicTetra-n-butylammonium phosphateTetrabutylammonium phosphate monobasicTetra-n-butylammonium dihydrogen phosphateIPC-TBA-P

AC43599-1000
103530-408EA 216.92 USD
103530-408
Tetrabutylammonium dihydrogen phosphate 0.4 M in acetonitrile, AcroSeal™
Tetrabutylammonium dihydrogen phosphate
Formula: C₁₆H₃₈NO₄P
MW: 339.46 g/mol
Boiling Pt: 81…82 °C (760 mmHg)
Melting Pt: –48 °C
Density: 0.828 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00064526
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to light yellow Solution
Titration with NaOH 0.38 to 0.45 M
Melting point 215 to 220 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR