4-Chlorotetrahydropyran 96%

Warning

AC43427-0010 AC43427-0050
103529-794EA 63.3 USD
103529-794 103529-796
4-Chlorotetrahydropyran 96%
4-Chlorotetrahydropyran
Formula: C₅H₉ClO
MW: 120.5800 g/mol
Boiling Pt: 150 °C (760 mmHg)
Density: 1.12 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00074911
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to dark yellow Liquid
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR