8-Quinolinesulfonyl chloride 98%

Danger

Synonyms: Quinoline-8-sulphonyl chlorideQuinoline-8-sulfonyl chloride

AC13221-0250 AC13221-0050
103530-938EA 271 USD
103530-938 103530-936
8-Quinolinesulfonyl chloride 98%
8-Quinolinesulphonyl chloride
Formula: C₉H₆ClNO₂S
MW: 227.67 g/mol
Boiling Pt: 306 °C (760 mmHg)
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00006808
CAS Number: 18704-37-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 130 °C
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR