5-Chloro-2-methoxypyridine 97%

Warning

AC45373-0010 AC45373-0050
103534-156EA 69.3 USD
103534-156 103534-158
5-Chloro-2-methoxypyridine 97%
5-Chloro-2-methoxypyridine
Formula: C₆H₆ClNO
MW: 143.5700 g/mol
Boiling Pt: 180…184 °C
Density: 1.193 g/cm³
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD06254388
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR