4-Methyl-2-pentyne 95%

Danger

AC38097-0050 AC38097-0010
103532-778EA 84.97 USD
103532-778 103532-776
4-Methyl-2-pentyne 95%
4-Methyl-2-pentyne
Formula: C₆H₁₀
Boiling Pt: 71…73 °C (760 mmHg)
Melting Pt: –110 °C
Density: 0.716 g/cm³
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00041615
CAS Number: 21020-27-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR