Tetramethyl orthocarbonate 99%

Danger

Synonyms: Tetramethylorthocarbonate

AC15811-0050 AC15811-0250
103531-472EA 116 USD
103531-472 103531-474
Tetramethyl orthocarbonate 99%
Tetramethyl orthocarbonate
Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 114 °C (760 mmHg)
Melting Pt: –5 °C
Density: 1.02 g/cm³
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00008473
CAS Number: 1850-14-2
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR