Ethyl-4,4,4-trifluorobutyrate 98%

Warning

Synonyms: Ethyl 4,4,4-trifluorobutanoateEthyl 4,4,4-trifluorobutyrate

AC30965-0050
103532-290EA 102 USD
103532-290
Ethyl-4,4,4-trifluorobutyrate 98%
Ethyl 4,4,4-trifluorobutyrat
Formula: C₆H₉F₃O₂
MW: 170.13 g/mol
Boiling Pt: 127 °C
Density: 1.16 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00041398
CAS Number: 371-26-6
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR