(±)-α-Bromophenylacetic acid 97%

Danger

Synonyms: α-Bromophenylacetic AcidDL-α-Bromophenylacetic acid

AC10727-0050 AC10727-0250 AC10727-1000
103530-250EA 256 USD
103530-250 103530-248 103530-246
(±)-α-Bromophenylacetic acid 97%
(±)-α-Bromophenylacetic acid
Formula: C₈H₇BrO₂
MW: 215.0440 g/mol
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00004206
CAS Number: 4870-65-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR