2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 97% (mixture of cis and trans)

Warning

Synonyms: 2,5-Dimethoxy-2,5-dihydrofuran

AC11553-1000
103530-564EA 92.5 USD
103530-564
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 97% (mixture of cis and trans)
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers)
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 160…162 °C (760 mmHg)
Density: 1.07 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00003220
CAS Number: 332-77-4
EINECS: 206-367-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR