Vanadium(III) chloride tetrahydrofuran complex (1:3) in methylene chloride, AcroSeal™

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Trichlorotris(tetrahydrofuran)vanadiumTrichlorotris(tetrahydrofuran)vanadium(III)Vanadium(III) chloride tetrahydrofuran adduct

AC43366-1000 AC43366-8000
103529-782EA 133.68 USD
103529-782 103529-784
Vanadium(III) chloride tetrahydrofuran complex (1:3) in methylene chloride, AcroSeal™
Vanadium(III) chloride tetrahydrofuran complex (1:3)
Formula: VCl₃·3C₄H₈O
Density: 1,312 g/cm³
Flash Pt: -8°C
Storage Temperature: Ambient
MDL Number: MFCD29050135
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light red to dark red Solution
Titration Argentometric 0.40 to 0.60 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR