5-Amino-1,4-benzodioxane 95%

Danger

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxin-5-amine1,4-Benzodioxan-5-amine

AC42937-0010 AC42937-0050
103533-230EA 586 USD
103533-230 103533-228
5-Amino-1,4-benzodioxane 95%
5-Amino-1,4-benzodioxane
Formula: C₈H₉NO₂
MW: 151.1626 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03695459
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear brown to purple Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Sulfite (SO3) ≤5 ppm
Sulfate (SO4) ≤5 ppm
Arsenic (As) ≤0.5 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR