1,3-Dichloroisoquinoline 97%
|
Warning
|
AC38241-0010
AC38241-0050
103532-800EA
223
USD
103532-800
103532-798
1,3-Dichloroisoquinoline 97%
1,3-Dichloroisoquinoline
Specification Test Results
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 120 to 125 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...