4-Bromophenyl phenyl ether (BDE No. 3) 99%

Danger

Synonyms: 1-Bromo-4-phenoxybenzene4-Bromodiphenyl ether

AC10689-0250 AC10689-0050
103530-240EA 49.5 USD
103530-240 103530-214
4-Bromophenyl phenyl ether (BDE No. 3) 99%
4-Bromophenyl phenyl ether (BDE No. 3)
Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Boiling Pt: 305 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1.423 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000094
CAS Number: 101-55-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.6065 to 1.6095 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR