8-Bromoisoquinoline 97%

Danger

AC45876-0050 AC45876-0010
103529-956EA 68.3 USD
103529-956 103529-958
8-Bromoisoquinoline 97%
8-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.0600 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Ambient
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 74 to 82 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR