Isoquinoline-8-amine 97%

Warning

Synonyms: Isoquinolin-8-amine8-Aminoisoquinoline

AC42940-0010 AC42940-0050
103533-232EA 210 USD
103533-232 103533-234
Isoquinoline-8-amine 97%
Isoquinoline-8-amine
Formula: C₉H₈N₂
MW: 144.1700 g/mol
Melting Pt: 168…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00179553
CAS Number: 23687-27-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 167 to 170 °C
GC ≥96.0 %
Residue after evaporation ≤0.0003 %
Conductivity ≤1 µS/cm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR