Bis(4-hydroxyphenyl) sulfide 99%

Danger

Synonyms: 4,4'-Thiodiphenol

AC20197-0250
103531-770EA 26.3 USD
103531-770
Bis(4-hydroxyphenyl) sulfide 99%
Bis(4-hydroxyphenyl) sulphide
Formula: C₁₂H₁₀O₂S
MW: 218.28 g/mol
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002349
CAS Number: 2664-63-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 155 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR