Lithium tetrachlorocuprate 0.1M in THF, AcroSeal™

Danger

Synonyms: Dilithium tetrachlorocuprate

AC42905-1000
103530-324EA 69.46 USD
103530-324
Lithium tetrachlorocuprate 0.1M in THF, AcroSeal™
Lithium tetrachlorocuprate
Formula: Cl₄CuLi₂
MW: 219.24 g/mol
Density: 0.91 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011081
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear orange to red-brown Solution
Titration Argentometric 0.09 to 0.12 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR