4-Aminopiperidine 95%

Danger

Synonyms: Piperidin-4-amine

AC43292-0250 AC43292-0050
103533-366EA 88.8 USD
103533-366 103533-368
4-Aminopiperidine 95%
4-Aminopiperidine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 65 °C (18Torr)
Density: 0.954 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD02179399
CAS Number: 13035-19-3
UN: 2733
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
GC ≥94.0 %
UV (0.5 M in water)
UV at 210 nm A: ≤0.1
UV at 220 nm A: ≤0.06
UV at 230 nm A: ≤0.04
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR