3-Piperidinemethanol 98%

Danger

Synonyms: 3-(Hydroxymethyl)piperidine

AC16804-1000 AC16804-0250 AC16804-0050
103531-612EA 39.8 USD
103531-612 103531-614 103531-616
3-Piperidinemethanol 98%
3-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 236 °C (760 mmHg)
Melting Pt: 58…62 °C
Density: 1.02 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00005997
CAS Number: 4606-65-9
UN: 3263
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR